Learn More
Hamilton™ Hitachi™ LaChrom™ Autosampler Syringes
Click to view available options
Volume (Metric):
500 μL
Unit Size:
Each
Description
- Provides improved reproducibility and speed
- For use with Hitachi™ LaChrom™ HPLC autosamplers
- Special termination
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Autosampler syringe |
| Volume (Metric) | 500 μL |
| For Use With (Equipment) | Hitachi LaChrom Autosamplers |
| Connection Type | Special |