missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Modafinil, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Modafinil
Specifications
Specifications
| Chemical Name or Material | Modafinil |
| CAS | 68693-11-8 |
| Analyte or Component Names | Modafinil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H15 N O2 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Modafinil, 2-[(RS)-(Diphenylmethyl)sulfinyl]acetamide, 2-(Benzhydrylsulfinyl)acetamide, CEP 1538, CRC 40476, CRL 40476, DEP 1538, Modaphonil, Modiodal, Provigil, Acetamide, 2-[(diphenylmethyl)sulfinyl]-(+/-)-Modafinil |
| InChI Formula | InChI=1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17) |
| SMILES | NC(=O)CS(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-benzhydrylsulfinylacetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction