missing translation for 'onlineSavingsMsg'
Learn More

Modafinil 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30458531
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30458531

Brand: LoGiCal LGCAMP0540.0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Modafinil
CAS 68693-11-8
Analyte or Component Names Modafinil
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H15 N O2 S
Certifications/Compliance ISO 17034
Synonym Modafinil, 2-[(RS)-(Diphenylmethyl)sulfinyl]acetamide, 2-(Benzhydrylsulfinyl)acetamide, CEP 1538, CRC 40476, CRL 40476, DEP 1538, Modaphonil, Modiodal, Provigil, Acetamide, 2-[(diphenylmethyl)sulfinyl]-(+/-)-Modafinil
InChI Formula InChI=1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)
Solution Type Single Solution
SMILES NC(=O)CS(=O)C(c1ccccc1)c2ccccc2
IUPAC Name 2-benzhydrylsulfinylacetamide
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 273.35
Quantity 1 mL
Formula Weight 273.0823 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.