missing translation for 'onlineSavingsMsg'
Learn More

MOCS3 Antibody (1C5-E8), Novus Biologicals™

Product Code. 18355529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355529 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355529 Supplier Novus Biologicals Supplier No. H00027304M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MOCS3 Monoclonal antibody specifically detects MOCS3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCS3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C5-E8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH15939
Gene Alias adenylyltransferase and sulfurtransferase MOCS3, dJ914P20.3, molybdenum cofactor synthesis 3, Molybdenum cofactor synthesis protein 3, Molybdopterin synthase sulfurylase, MPT synthase sulfurylase, UBA4, ubiquitin-activating enzyme E1 homolog, UBA4MGC9252, ubiquitin-like modifier activating enzyme 4
Host Species Mouse
Immunogen MOCS3 (AAH15939, 1 a.a. ~ 460 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASREEVLALQAEVAQREEELNSLKQKLASALLAEQEPQPERLVPVSPLPPKAALSRDEILRYSRQLVLPELGVHGQLRLGTACVLIVGCGGLGCPLAQYLAAAGVGRLGLVDYDVVEMSNLARQVLHGEALAGQAKAFSAAASLRRLNSAVECVPYTQALTPATALDLVRRYDVVADCSDNVPTRYLVNDACVLAGRPLVSASALRFEGQITVYHYDGGPCYRCIFPQPPPAETVTNCADGGVLGVVTGVLGCLQALEVLKIAAGLGPSYSGSLLLFDALRGHFRSIRLRSRRLDCAACGERPTVTDLLDYEAFCGSSATDKCRSLQLLSPEERVSVTDYKRLLDSGAFHLLLDVRPQVEVDICRLPHALHIPLKHLERRDAESLKLLKEAIWEEKQGTQEGAAVPIYVICKLGNDSQKAVKILQSLSAAQELDPLTVRDVVGGLMAWAAKIDGTFPQY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27304
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.