missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOCS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14416-1-AP-20UL
This item is not returnable.
View return policy
Description
Eukaryotic molybdoenzymes use a unique molybdenum cofactor (MoCo) consisting of a pterin, termed molybdopterin, and the catalytically active metal molybdenum. MoCo is synthesized from precursor Z by the heterodimeric enzyme molybdopterin synthase. The large and small subunits of molybdopterin synthase are both encoded from this gene by overlapping open reading frames. The proteins were initially thought to be encoded from a bicistronic transcript. They are now thought to be encoded from monocistronic transcripts. Alternatively spliced transcripts have been found for this locus that encode the large and small subunits.Specifications
| MOCS2 | |
| Polyclonal | |
| Unconjugated | |
| MOCS2 | |
| MCBPE, MOCO1, MOCO1 A, MOCS2, MOCS2A, MOCS2B, MPTS, Sulfur carrier protein MOCS2A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17434, 294753, 4338 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O96033, Q6AY59, Q9Z224 | |
| Mocs2 | |
| MOCS2 Fusion Protein Ag5761 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction