missing translation for 'onlineSavingsMsg'
Learn More

MOCS2 Antibody (4H3), Novus Biologicals™

Product Code. 18388477 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388477 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388477 Supplier Novus Biologicals Supplier No. H00004338M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MOCS2 Monoclonal antibody specifically detects MOCS2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCS2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4H3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_789776.1
Gene Alias EC 2.-, MCBPE, MOCO1-A, MOCO1-B, MOCO1MOCS2B, MOCS2A, molybdenum cofactor biosynthesis protein E, molybdenum cofactor synthesis 2, Molybdenum cofactor synthesis protein 2 large subunit, Molybdenum cofactor synthesis protein 2 small subunit, Molybdenum cofactor synthesis protein 2A, Molybdenum cofactor synthesis protein 2B, molybdopterin synthase catalytic subunit, molybdopterin synthase sulfur carrier subunit, Molybdopterin-synthase large subunit, Molybdopterin-synthase small subunit, MPT synthase large subunit, MPTS, Sulfur carrier protein MOCS2A
Host Species Mouse
Immunogen MOCS2 (NP_789776.1, 1 a.a. ~ 88 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVPLCQVEVLYFAKSAEITGVRSETISVPQEIKALQLWKEIETRHPGLADVRNQIIFAVRQEYVELGDQLLVLQPGDEIAVIPPISGG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4338
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.