missing translation for 'onlineSavingsMsg'
Learn More

Moclobemide, TRC

Product Code. 30137309
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137309 50 mg 50mg
30135488 25 mg 25mg
17922175 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137309 Supplier Toronto Research Chemicals Supplier No. TRCM48100050MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13 H17 Cl N2 O2
Molecular Weight (g/mol) 268.74
Synonym Moclobemide, Benzamide, 4-chloro-N-[2-(4-morpholinyl)ethyl]- (9CI, ACI), 4-Chloro-N-[2-(4-morpholinyl)ethyl]benzamide (ACI), 4-Chloro-N-(2-morpholin-4-ylethyl)benzamide, 4-Chloro-N-(2-morpholinoethyl)benzamide, Auromid, Aurorix, Manerix, MeSH ID: D020912, Moclaime, Moclobemide, p-Chloro-N-(2-morpholinoethyl)-benzamide, Ro 11-1163, Ro 11-1163/000
IUPAC Name 4-chloro-N-(2-morpholin-4-ylethyl)benzamide
SMILES Clc1ccc(cc1)C(=O)NCCN2CCOCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H17ClN2O2/c14-12-3-1-11(2-4-12)13(17)15-5-6-16-7-9-18-10-8-16/h1-4H,5-10H2,(H,15,17)
Formula Weight 268.0979
Percent Purity >95%
Chemical Name or Material Moclobemide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.