missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Moclobemide
GREENER_CHOICE

Product Code. 18792694
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792694 10 mg 10mg
18742494 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18792694 Supplier Tocris Bioscience™ Supplier No. 4395/10
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Reversible MAO-A inhibitor

Reversible monoamine oxidase A (MOA-A) inhibitor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13H17ClN2O2

Specifications

Quantity 10 mg
Formula Weight 268.74
Purity >99%
Molecular Weight (g/mol) 268.741
Chemical Name or Material Moclobemide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.