missing translation for 'onlineSavingsMsg'
Learn More

Moclobemide, MedChemExpress

Product Code. 16444387
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16444387

Brand: Medchem Express HYB053450mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Moclobemide (Ro111163) is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A.Moclobemide up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice.

Moclobemide (Ro111163) is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A.Moclobemide up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice.

Specifications

Chemical Name or Material Moclobemide
Physical Form Solid
CAS 71320-77-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H17ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Ro111163
Solubility Information DMSO : 100 mg/mL (372.11 mM; Need ultrasonic)
SMILES O=C(NCCN1CCOCC1)C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 268.74
Formula Weight 268.74
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.