missing translation for 'onlineSavingsMsg'
Learn More

MOBKL2B Antibody (OTI6C6), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-72748CL1

Product Code. 30145592

  • £870.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MOBKL2B Monoclonal antibody specifically detects MOBKL2B in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MOBKL2B
Monoclonal
CoraFluor 1
Em:AL163192.1, FLJ13204, FLJ23916, Mob1 homolog 2b, MOB1, Mps One Binder kinase activator-like 2B (yeast), MOB3BMGC32960, monopolar spindle 1 binding, MOB1, domain containing, mps one binder kinase activator-like 2B, Protein Mob3B
Full length human recombinant protein of human MOBKL2B (NP_079037) produced in HEK293T cell.
0.1 mL
Primary
Human, Mouse, Rat
Purified
Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence
OTI6C6
PBS
Mouse
Immunogen affinity purified
RUO
79817
Store at 4°C in the dark. Do not freeze.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.