missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOBKL1B Antibody (2D3D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61873-0.025ml
This item is not returnable.
View return policy
Description
MOBKL1B Monoclonal specifically detects MOBKL1B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| MOBKL1B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| C2orf6, chromosome 2 open reading frame 6, FLJ10788, FLJ11595, MATS1, MOB1, Mob1 alpha, Mob1 homolog 1B, MOB1, Mps One Binder kinase activator-like 1B (yeast), mob1A, Mob4B, MOBK1B, mps one binder kinase activator-like 1B, Protein Mob4B | |
| Mouse | |
| 25 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 2D3D11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MOB1A | |
| Purified recombinant fragment of human MOB1A (AA: 1-216) expressed in E. Coli. | |
| Protein G purified | |
| Cancer | |
| 55233 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction