missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ MobileALERT for UN75plus
Shop All Memmert Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
UN75plus Oven
missing translation for 'unitSize'
Each
Description
- MobileALERT
- Notification by SMS in case of any error or alarm of the device
- Requires option H6
Specifications
Specifications
| For Use With (Equipment) | UN75plus Oven |
| Type | Alerting Device |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction