Learn More
Miele Professional™ Mobile Unit A 200
Product Code. 15437704 Shop All Miele Products
Change view
Click to view available options
Dimensions (D x W x H): 546 x 529 x 357mm
Product Code.
15437704 Dimensions (D x W x H): 546 x 529 x 357mm
1 options
Product Code. 15437704 Supplier Miele Professional™ Supplier No. 12667060
  • ONLINE EXCLUSIVE PRICE
    £2699.42
    £2190.00 / Pack of 1
    Save £509.42
    18% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
Mobile unit A 200 for the optimum loading of 38 pipettes
  • For 38 pipettes in three rows
  • 1st row: 10 pipettes, 100mL, holder spacing 20mm
  • 2nd row: 14 pipettes, 25mL, holder spacing 26mm
  • 3rd row: 14 pipettes, 10mL, holder spacing 26mm
  • H 397mm, W 529mm, D 546mm