missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Mobile Unit

Product Code. 15410575 Shop All Miele Products
This item is not returnable. View return policy
Product Code. 15410575 Supplier Miele Professional™ Supplier No. 12667130
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Mobile unit A 202 for the optimum loading of 4 DIN mesh trays

  • Suitable for 4 DIN mesh trays on 2 levels
  • Insert frames: Removable for the reprocessing of tall wash items
  • 6 additional connection options via Luer lock connection
  • With integrated spray arm
  • Loading volumes for the different levels differ from each other
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Product Type Mobile Unit for Mesh Trays
Color Stainless Steel
Holds 4 DIN mesh trays
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 542 mm
Height (Metric) 223 mm
Width (Metric) 529 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.