missing translation for 'onlineSavingsMsg'
Learn More

Mobile Phase: Hexane/Ethyl Ether/Isopropyl Alcohol, Ricca

Product Code. 31940763
Change view
Click to view available options
Packaging:
Amber Glass Container
Safety-coated Glass Container
Unit Size:
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
31940763 Amber Glass Container 4 L
31940645 Safety-coated Glass Container 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31940763 Supplier Ricca Chemical Company Supplier No. R51811004C
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Mobile Phase Hexane/Ether/IPA
  • Available in 4 L packaging in amber glass containers

Specifications

Form Liquid
Reagent Type Solvent Mixture
For Use With Mobile Phase Eluent
Packaging Amber Glass Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.