Learn More
Manutan Mobile ingredient truck, blue, 90 litre capacity
Product Code. 12834269
Encompass_Preferred
Product Code. 12834269 Supplier Manutan Supplier No. A338873
Only null left
Add to basket
Add to basket
Suitable for storing and transporting all types of ingredients, ideal for food environments, with a clear lid for view of contents. Tough, 100% food grade medium density polyethylene, easy to clean. Polycarbonate shatter proof lids fitted. Four x 76mm swivel castors fitted as standard.
  • Capacity: 90L
  • Dimensions (H x W x D): 660mm x 470mm x 610mm
  • Internal dimensions (H x W x D): 565mm x 385mm x 535mm