missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOB4A/MOBKL1B Antibody (01), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | MOB4A/MOBKL1B |
|---|---|
| Clone | 01 |
| Dilution | Immunocytochemistry/ Immunofluorescence 1:20-1:100 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684469
|
Novus Biologicals
NBP3-06512-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632809
|
Novus Biologicals
NBP3-06512-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MOB4A/MOBKL1B Monoclonal antibody specifically detects MOB4A/MOBKL1B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| MOB4A/MOBKL1B | |
| Immunocytochemistry/ Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 92597 | |
| IgG2a | |
| Protein A purified |
| 01 | |
| Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| ATS2;MATS2;MOB kinase activator 1B;MOB1B;MOB4A;MOBKL1A | |
| This antibody was obtained from a mouse immunized with purified, recombinant Human MOB4A/MOBKL1B (Uniprot#: Q7L9L4; Met1-Arg216). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title