missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOB4A Antibody (OTI3G9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72744CL1
This item is not returnable.
View return policy
Description
MOB4A Monoclonal antibody specifically detects MOB4A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| MOB4A | |
| Monoclonal | |
| CoraFluor 1 | |
| MATS2, MGC33910, Mob1 homolog 1A, MOB1, Mps One Binder kinase activator-like 1A (yeast), Mob1A, Mob1Bmps one binder kinase activator-like 1A, MOB4Amob1A, Protein Mob4A | |
| Full length human recombinant protein of human MOBKL1A(NP_775739) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 92597 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| OTI3G9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction