missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MOB4A Polyclonal antibody specifically detects MOB4A in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | MOB4A |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | MATS2, MGC33910, Mob1 homolog 1A, MOB1, Mps One Binder kinase activator-like 1A (yeast), Mob1A, Mob1Bmps one binder kinase activator-like 1A, MOB4Amob1A, Protein Mob4A |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human MOB4A (Uniprot#: Q7L9L4; Met1-Arg216) |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?