missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551232
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MOB4 protein.
This gene was identified based on its similarity with the mouse counterpart. Studies of the mouse counterpart suggest that the expression of this gene may be regulated during oocyte maturation and preimplantation following zygotic gene activation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. Naturally occurring read-through transcription occurs between this locus and the neighboring locus HSPE1.
Specifications
| MOB4 | |
| Polyclonal | |
| Unconjugated | |
| MOB4 | |
| 2610109B12Rik; 2C4D; CGI-95; Class II mMOB1; MOB family member 4, phocein; MOB1; Mob1 homolog 3; MOB1, Mps One Binder kinase activator-like 3; mob3; MOB4; Mobkl1; Mobkl3; MOB-like protein phocein; mps one binder kinase activator-like 3; Phocein; phocein, Mob-like protein; PHOCN; PREI3; Preimplantation protein 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171050, 19070, 25843 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6PEB6, Q9QYW3, Q9Y3A3 | |
| MOB4 | |
| Full length fusion protein of human MOB4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction