missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOB3C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538715
This item is not returnable.
View return policy
Description
The protein encoded by this gene is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq, Jul 2008].
Specifications
| MOB3C | |
| Polyclonal | |
| Unconjugated | |
| MOB3C | |
| MOB1E, MOBKL2C, MOB1, Mps One Binder kinase activator-like 2C, mob1 homolog 2C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100465, 148932 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q70IA8, Q8BJG4 | |
| MOB3C | |
| A synthetic peptide derived from the internal region of human MOL2C | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction