missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOB3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104364
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MOBKL2B.
The protein encoded by this gene shares similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. This gene is located on the opposite strand as the interferon kappa precursor (IFNK) gene.
Specifications
| MOB3B | |
| Polyclonal | |
| Unconjugated | |
| MOB3B | |
| 8430436F23Rik; A430018A01Rik; C9orf35; MOB kinase activator 3B; MOB kinase activator-like 2B; MOB1 (Mps One Binder) kinase activator-like 2B; mob1 homolog 2b; MOB1, Mps One Binder kinase activator-like 2B; MOB1D; MOB3B; MOBKL2B; monopolar spindle 1 binding, MOB1, domain containing; mps one binder kinase activator-like 2B; RGD1560696 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 214944, 79817 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86TA1, Q8VE04 | |
| MOB3B | |
| A synthesized peptide derived from human MOB3B(Accession Q86TA1), corresponding to amino acid residues A180-H216. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction