missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOB1A Monoclonal Antibody (2D3D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531800
This item is not returnable.
View return policy
Description
MA5-31800 has been tested in indirect ELISA.
The protein encoded by this gene is a component of the Hippo signaling pathway, which controls organ size and tumor growth by enhancing apoptosis. Loss of the encoded protein results in cell proliferation and cancer formation. The encoded protein is also involved in the control of microtubule stability during cytokinesis. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| MOB1A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9H8S9 | |
| MOB1A | |
| Purified recombinant fragment of human MOB1A (AA: 1-216) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 2D3D11 | |
| Unconjugated | |
| MOB1A | |
| C2orf6; MATS1; MATS2; MOB kinase activator 1A; MOB kinase activator 1B; MOB1; Mob1 alpha; mob1 homolog 1A; Mob1 homolog 1B; MOB1 Mps One Binder homolog A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; MOB1, Mps One Binder kinase activator-like 1B; mob1A; MOB1B; MOB4A; Mob4B; MOBK1B; MOBKL1A; MOBKL1B; Mps one binder kinase activator-like 1A; mps one binder kinase activator-like 1B | |
| Mouse | |
| Protein G | |
| RUO | |
| 55233 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction