missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MO-I-500
GREENER_CHOICE

Product Code. 18767216
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18767216

Brand: Tocris Bioscience™ 6871/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FTO inhibitor; anticonvulsant

MO-I-500 is a FTO inhibitor (IC50 = 8.7 μM for purified FTO in vitro), which increases N6-methyladenosine (m6A) levels in total cellular mRNA in HeLa cells. MO-I-500 modulates levels of specific miRNAs. In a triple-negative breast cancer cell line metabolically challenged by culture without glutamine (SUM149-MA cells), MO-I-500 inhibits cell proliferation. MO-I-500 also displays anticonvulsant activity in a 6 Hz mouse model of epilepsy at nontoxic doses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[5-(4-Chlorophenyl)-3,4-dihydroxy-2-furanyl]ethanesulfonamide
CAS 1585219-04-0
Quantity 10 mg
Target Other Oxidase/Oxygenase Compounds
Molecular Formula C12H12ClNO5S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.