missing translation for 'onlineSavingsMsg'
Learn More

MNX1/HLXB9 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18426617
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426617 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426617 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224691MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MNX1/HLXB9 Polyclonal antibody specifically detects MNX1/HLXB9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNX1/HLXB9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias HB9, HLXB9motor neuron and pancreas homeobox protein 1, homeobox HB9, HOXHB9Homeobox protein HB9, motor neuron and pancreas homeobox 1, SCRA1homeo box HB9
Host Species Rabbit
Immunogen A portion of amino acids 330-380 of mouse MNX1/HLXB9 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3110
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.