missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNX1/HLXB9 Antibody (JE32-08), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32417
This item is not returnable.
View return policy
Description
MNX1/HLXB9 Monoclonal antibody specifically detects MNX1/HLXB9 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| MNX1/HLXB9 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| HB9, HLXB9motor neuron and pancreas homeobox protein 1, homeobox HB9, HOXHB9Homeobox protein HB9, motor neuron and pancreas homeobox 1, SCRA1homeo box HB9 | |
| Synthetic peptide within Human MNX1/HLXB9 aa 276-325/ 401. (Uniprot: P50219) | |
| 100 μL | |
| Cell Biology, Neuroscience, Stem Cells | |
| 3110 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE32-08 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction