missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12693-1-AP-20UL
This item is not returnable.
View return policy
Description
MNS1 may play a role in the control of meiotic division and germ cell differentiation through regulation of pairing and recombination during meiosis.Specifications
| MNS1 | |
| Polyclonal | |
| Unconjugated | |
| MNS1 | |
| meiosis nuclear structural 1, MNS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17427, 363093, 55329 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q61884, Q6AXQ8, Q8NEH6 | |
| Mns1 | |
| MNS1 Fusion Protein Ag3423 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction