missing translation for 'onlineSavingsMsg'
Learn More

MNS, MedChemExpress

Product Code. 16481956
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16481956

Brand: Medchem Express HY78263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MNS (NSC 170724), the beta-nitrostyrene derivative, is a potent tyrosine kinase inhibitor and a broad-spectrum antiplatelet agent. MNS completely inhibits U46619, ADP-, arachidonic acid-, collagen-, and thrombin-induced platelet aggregation with IC50 values of 2.1, 4.1, 5.8, 7.0, and 12.7 μM, respectively. MNS inhibits Src, Syk, and FAK with IC50 of 27.3, 2.8, and 97.6 μM, respectively.

MNS (NSC 170724), the beta-nitrostyrene derivative, is a potent tyrosine kinase inhibitor and a broad-spectrum antiplatelet agent. MNS completely inhibits U46619, ADP-, arachidonic acid-, collagen-, and thrombin-induced platelet aggregation with IC50 values of 2.1, 4.1, 5.8, 7.0, and 12.7 μM, respectively. MNS inhibits Src, Syk, and FAK with IC50 of 27.3, 2.8, and 97.6 μM, respectively.

Specifications

Chemical Name or Material MNS
CAS 1485-00-3
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C9H7NO4
Quantity 500 mg
Synonym NSC 170724 5-(2-Nitrovinyl)benzodioxole
Solubility Information DMSO : 50 mg/mL (258.85 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (3.47 mM; Need ultrasonic)
SMILES O=[N+]([O-])/C=C/C1=CC(OCO2)=C2C=C1
Molecular Weight (g/mol) 193.16
Formula Weight 193.16
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.