missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58899
This item is not returnable.
View return policy
Description
MNK2 Polyclonal specifically detects MNK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MNK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, G protein-coupled receptor kinase 7, GPRK7Putative map kinase interacting kinase, MAP kinase interacting serine/threonine kinase 2, MAP kinase signal-integrating kinase 2, Mnk2, MNK2MAP kinase-interacting serine/threonine-protein kinase 2 | |
| Rabbit | |
| 51 kDa | |
| 100 μL | |
| Protein Kinase | |
| 2872 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9HBH9 | |
| MKNK2 | |
| Synthetic peptides corresponding to MKNK2(MAP kinase interacting serine/threonine kinase 2) The peptide sequence was selected from the N terminal of MKNK2. Peptide sequence SDFGLQCSARPDMPASQPIDIPDAKKRGKKKKRGRATDSFSGRFEDVYQL. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 100%; Bovine: 92%; Xenopus: 83%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction