missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MNI 137
GREENER_CHOICE

Product Code. 30001507
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001507 10 mg 10mg
30001472 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001507 Supplier Tocris Bioscience™ Supplier No. 4388/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective negative allosteric modulator of group II mGlu receptors

Selective negative allosteric modulator of group II mGlu receptors (IC50 values are 8.3 and 12.6 nM for human and rat mGlu2 inhibition of glutamate-induced calcium mobilization). Displays no activity at mGlu1, mGlu4, mGlu5 or mGlu8 receptors in a calcium mobilization assay.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946619-21-2
Molecular Formula C15H9BrN4O

Specifications

Quantity 10 mg
Formula Weight 341.16
Purity >98%
Molecular Weight (g/mol) 341.168
Chemical Name or Material MNI 137
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.