Learn More
mNeonGreen Mouse, Unconjugated, Clone: 32F6, Proteintech ChromoTek
Mouse Monoclonal Antibody
Brand: Chromotek 32F6-100
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody [32F6] to mNeonGreen fluorescent protein derived from Branchiostoma lanceolatum.
Specifications
| mNeonGreen | |
| Monoclonal | |
| Unconjugated | |
| 100 μL | |
| Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| Western Blot, Immunofluorescence, ELISA | |
| 32F6 | |
| Mouse | |
| Primary | |
| IgG2c |