missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MNDA Antibody Blocking Peptide

Product Code. 18368312
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368312 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368312 Supplier Novus Biologicals™ Supplier No. NBP318135PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blocking Peptied

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/ml
For Use With (Application) Antibody Competition
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin
Gene ID (Entrez) 4332
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias myeloid cell nuclear differentiation antigen, PYHIN3
Gene Symbol MNDA
Research Category DNA replication Transcription Translation and Splicing
Protein MNDA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.