missing translation for 'onlineSavingsMsg'
Learn More

MNDA Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18349016 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349016 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349016 Supplier Novus Biologicals Supplier No. NBP318135AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MNDA Polyclonal antibody specifically detects MNDA in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNDA
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias myeloid cell nuclear differentiation antigen, PYHIN3
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the center of human MNDA. The immunogen is located within 110-160 amino acids of MNDA.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 4332
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.