missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MND1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11636-1-AP-150UL
This item is not returnable.
View return policy
Description
Required for proper homologous chromosome pairing and efficient cross-over and intragenic recombination during meiosis. Stimulates both DMC1- and RAD51-mediated homologous strand assimilation, which is required for the resolution of meiotic double-strand breaks.Specifications
| MND1 | |
| Polyclonal | |
| Unconjugated | |
| MND1 | |
| GAJ, MND1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 76915, 84057 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K396, Q9BWT6 | |
| Mnd1 | |
| MND1 Fusion Protein Ag2210 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction