missing translation for 'onlineSavingsMsg'
Learn More

MNAT1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18362582 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362582 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362582 Supplier Novus Biologicals Supplier No. NBP306477DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MNAT1 Polyclonal antibody specifically detects MNAT1 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNAT1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CAP35, CDK7/cyclin-H assembly factor, cyclin G1 interacting protein, Cyclin-G1-interacting protein, MAT1TFB3, Menage a trois, menage a trois 1 (CAK assembly factor), menage a trois homolog 1, cyclin H assembly factor (Xenopus laevis), p35, p36, RING finger protein 66, RING finger protein MAT1, RNF66CDK-activating kinase assembly factor MAT1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MNAT1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4331
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.