missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MNAR Recombinant Rabbit Monoclonal Antibody (24GB1145)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557555
This item is not returnable.
View return policy
Description
MNAR Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications
| MNAR | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PELP1 | |
| 4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27043, 360552, 75273 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1145 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q56B11, Q8IZL8, Q9DBD5 | |
| PELP1 | |
| A synthesized peptide derived from human PELP1 (aa 1030-1065). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction