missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MNAR Monoclonal Antibody (2D6D11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531836
This item is not returnable.
View return policy
Description
MA5-31836 has been tested in indirect ELISA.
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications
| MNAR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q8IZL8 | |
| PELP1 | |
| Purified recombinant fragment of human PELP1 (AA: 1031-1180) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Western Blot | |
| 2D6D11 | |
| Unconjugated | |
| PELP1 | |
| 4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 27043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction