missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MN1 Polyclonal antibody specifically detects MN1 in Human samples. It is validated for ELISA,Immunohistochemistry
Specifications
Specifications
| Antigen | MN1 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | dJ353E16.2, meningioma (disrupted in balanced translocation) 1, meningioma (translocation balanced), meningioma chromosome region, meningioma chromosome region 1, MGCR, MGCR1, MGCR1-PEN, probable tumor suppressor protein MN1 |
| Host Species | Rabbit |
| Immunogen | meningioma(disrupted in balanced translocation) 1. (Uniprot# Q10571) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?