missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ MN Square-well Block

Product Code. 15800948
Click to view available options
Color:
White
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 15800948

Brand: Macherey-Nagel™ 740678

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A 96-well block with 2.2 mL square wells for use in processing NucleoSpin™ 8-well strips under centrifugation (e.g. NucleoSpin 8 Blood QuickPure, 8 Virus) as a waste or flow-through collection device. MN Square-well Block is included in Starter Set C (man. part no. 740684).

Starter Set C

  • Required for fist time users of NucleoSpin™ 8 kits
  • Contains: Column Holders C, MN Square-well Blocks for waste collection, and Rack of Tube Strips
  • For use of 8-well strips under centrifugation, using a microtiterplate centrifuge
    • Centrifuge must be able to accommodate an MN Square-well Block :
    • Centrifuge must be able to reach accelerations of 5,600 x g 
    • E.g. Hermle Z513, Qiagen/Sigma 4-15c, Jouan KR4i, KendroHeraeus Multifuge 3/3-R, Highplate™, Beckman Coulter, Allegra R25

Specifications

Color White
Product Type Square-well Block
For Use With (Equipment) NucleoSpin 8 kits; Centrifuge
Volume (Metric) Well 2.2 mL
No. of Wells 96
For Use With (Application) Processing 8-well strips under centrifugation
Includes Lid
Quantity 20 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.