missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ MN Science Silicon Septa
Click to view available options
Diameter (Metric) Inner:
11.5 mm
12.5 mm
17 mm
Unit Size:
Pack of 48
Description
- High mechanical strength septa - up to 400 injections per septa
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Specifications
| Type | MN |
| Diameter (Metric) Inner | 11.5 mm |
| Quantity | 48/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction