missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ MN Bead Tube Holder

Product Code. 15513626
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15513626

Brand: Macherey-Nagel™ 740469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubber-foam adapter for processing MN Bead Tubes with Vortex-Genie 2.

NucleoSpin™ Bead Tubes

  • Bead tubes for efficient sample homogenization of hard-to-lyse sample solutions
  • Provided in different variants (e.g., glass beads, corundum, steel beads)
  • Variants are appropriate for specific applications and/or sample material
  • Up to 60% more RNA and DNA compared to standard extraction methods

Specifications

Color Blue
Content And Storage 1 Ea., 15°C to 25°C
Format Horizontal
Isolation Technology Beads
For Use With (Equipment) Vortex-Genie™ instrument
For Use With (Application) Homogenization of hard-to-lyse samples
Product Type Tube Holder
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.