missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMS22L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566891
This item is not returnable.
View return policy
Description
Immunogen sequence: MSQVVPFSQLA DAAADFTLLA MDMPSTAPSD FQPQPVISII QLFGWDDIIC PQVVARYLSH VLQNSTLCEA LSHSGYVSFQ ALTVRSWIRC Highest antigen sequence identity to the following orthologs - mouse 81%, rat 82%.
Component of the MMS22L-TONSL complex, a complex that stimulates the recombination-dependent repair of stalled or collapsed replication forks. The MMS22L-TONSL complex is required to maintain genome integrity during DNA replication by promoting homologous recombination-mediated repair of replication fork-associated double-strand breaks. It may act by mediating the assembly of RAD51 filaments on ssDNA.
Specifications
| MMS22L | |
| Polyclonal | |
| Unconjugated | |
| MMS22L | |
| C6orf167; dJ39B17.2; Methyl methanesulfonate-sensitivity protein 22-like; MMS22 like, DNA repair protein; MMS22L; MMS22-like, DNA repair protein; Protein MMS22-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 253714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZRQ5 | |
| MMS22L | |
| Recombinant Human MMS22L. Recombinant protein control fragment (Product #RP-107538). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction