missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMRN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557392
This item is not returnable.
View return policy
Description
Immunogen sequence: RHNLLRNEVQ GRDDALERRI NEYALEMEDG LNKTMTIINN AIDFIQDNYA LKETLSTIKD NSEIHHKCTS DMETILTFIP QFHRLND Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 63%.
Multimerin-1, also known as MMRN1, EMILIN-4 or ECM (endothelial cell multimerin), is a 1,228 amino acid secreted protein that contains one C1q domain, one EMI domain and one EGF-like domain. Synthesized in megakaryocytes and endothelial cells and present in liver, lung and placenta, Multimerin-1 exists as a multimeric structure composed of varying disulfide-linked multimers and functions as a carrier protein for platelet factors (specifically platelet factor V), playing a role in the stabilization and storage of factor V in platelets. In addition, Multimerin-1 acts as a ligand for select Integrins and may participate in extracellular matrix adhesion. Defects in the gene encoding Multimerin-1 that lead to Multimerin-1 deficiency are associated with autosomal dominant bleeding disorders due to platelet factor malfunction. Multiple isoforms of Multimerin-1 exist due to alternative splicing events.
Specifications
| MMRN1 | |
| Polyclonal | |
| Unconjugated | |
| MMRN1 | |
| 155 kDa platelet multimerin; 4921530G03Rik; ECM; Elastin microfibril interface located protein 4; elastin microfibril interfacer 4; EMILIN4; EMILIN-4; Endothelial cell multimerin; glycoprotein Ia*; GPIa*; MMRN; mmrn1; multimerin 1; multimerin-1; p155; p-155; Platelet glycoprotein Ia* | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13201 | |
| MMRN1 | |
| Recombinant protein corresponding to Human MMRN1. Recombinant protein control fragment (Product #RP-96663). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction