missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMR/CD206/Mannose Receptor Rabbit anti-Human, Clone: S06-5I1, Novus Biologicals™
Rabbit Monoclonal Antibody
£232.00 - £451.00
Specifications
| Antigen | MMR/CD206/Mannose Receptor |
|---|---|
| Clone | S06-5I1 |
| Dilution | Western Blot |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314687
|
Novus Biologicals
NBP3-14900-25UL |
25 μg |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18370015
|
Novus Biologicals
NBP3-14900-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Western BlotSpecifications
| MMR/CD206/Mannose Receptor | |
| Western Blot | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen | |
| A synthetic peptide of human MMR/CD206/Mannose Receptor (Uniprot#: P22897) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-5I1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 4360 | |
| IgG | |
| Affinity purified | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title