missing translation for 'onlineSavingsMsg'
Learn More

MMR/CD206/Mannose Receptor Antibody (309210) [Biotin], Novus Biologicals™

Product Code. 30735778 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735778 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735778 Supplier Novus Biologicals Supplier No. FAB2534B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMR/CD206/Mannose Receptor
Applications Immunocytochemistry
Classification Monoclonal
Clone 309210
Conjugate Biotin
Formulation PBS
Gene Alias CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMR, Leu19-Lys1383 (Thr399Ala) & (Leu407Phe), Accession # P22897
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4360
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.