missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMR/CD206/Mannose Receptor Antibody (15-2) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional.
Specifications
Specifications
| Antigen | MMR/CD206/Mannose Receptor |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
| Classification | Monoclonal |
| Clone | 15-2 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry 1-4 ug/ml, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional |
| Formulation | PBS, 0.2 um filter sterilized |
| Gene Alias | CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?