missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MMPX
PDE1 inhibitor
Specifications
| Chemical Name or Material | 8-Methoxymethyl-3-isobutyl-1-methylxanthine |
|---|---|
| CAS | 78033-08-6 |
Description
Specific inhibitor of calmodulin-sensitive cyclic GMP phosphodiesterase (IC50 = 5.2 μM).Specifications
| 8-Methoxymethyl-3-isobutyl-1-methylxanthine | |
| 78033-08-6 |
Safety and Handling
Recommended Storage : Store at Room Temperature
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title