Learn More
Mmp9 Rabbit, anti-Mouse, Clone: 5A1, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82854-1-RR-20UL
This item is not returnable.
View return policy
Description
The Mmp9 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of mouse Mmp9. This antibody recognizes mouse antigen. The Mmp9 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
| Mmp9 | |
| 5A1 | |
| AW743869, B/MMP9, Clg4b, matrix metallopeptidase 9, MMP 9, Mmp9, pro MMP 9 | |
| 20 μL | |
| 17395 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse | |
| 150 kDa | |
| IgG |