missing translation for 'onlineSavingsMsg'
Learn More

MMP9 (N-terminal) Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech

Product Code. 17521666 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17521666 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17521666 Supplier Proteintech Supplier No. CL59410375100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The MMP9 (N-terminal) antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human MMP9 (N-terminal). This antibody recognizes human, mouse, rat antigen. The MMP9 (N-terminal) antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.

Specifications

Antigen MMP9 (N-terminal)
Applications Immunofluorescence
Classification Polyclonal
Conjugate CoraLite 594
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 4318
Target Species Human, Mouse, Rat
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.