missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551003
This item is not returnable.
View return policy
Description
MMP8 Polyclonal Antibody detects endogenous levels of MMP8 protein.
MMP8 Polyclonal antibody specifically detects MMP8 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Specifications
| MMP8 | |
| Polyclonal | |
| Unconjugated | |
| MMP8 | |
| BB138268; CLG1; Collagenase; Collagenase 2; collagenase-2; HNC; matrix metallo protease; matrix metallopeptidase 8; matrix metallopeptidase 8 (neutrophil collagenase); matrix metalloproteinase 8; matrix metalloproteinase 8 (neutrophil collagenase); Matrix metalloproteinase8; matrix metalloproteinase-8; MMP; MMP 8; Mmp8; MMP-8; MMPs; Neutrophil collagenase; neutrophil collagenease; PMN leukocyte collagenase; PMNL CL; PMNL collagenase; PMNLCL; PMNL-CL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17394, 4317, 63849 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70138, O88766, P22894 | |
| MMP8 | |
| Synthesized peptide derived from the C-terminal region of human MMP-8. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction